1-(3-Ethoxy-5-methylphenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

446919-5G 446919-25G 446919-1G 446919-500MG
77629-358EA 1583.11 USD
77629-358 77655-768 77642-860 77646-850
1-(3-Ethoxy-5-methylphenyl)cyclopentanol ≥95%
1-(3-Ethoxy-5-methylphenyl)cyclopentanol

Precaución: For Research Use Only

Formula: C₁₄H₂₀O₂
Storage Temperature: Ambient
CAS Number: 3031788-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR