1-(3-Ethoxy-5-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

446918-5G 446918-25G 446918-1G 446918-500MG
77629-356EA 1615.04 USD
77629-356 77659-360 77642-858 77646-848
1-(3-Ethoxy-5-methylphenyl)cyclobutanol ≥95%
1-(3-Ethoxy-5-methylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 206.281 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031793-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR