Cyclobutyl(3-ethoxy-5-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

446912-5G 446912-1G 446912-500MG
77629-354EA 1599.59 USD
77629-354 77642-856 77650-818
Cyclobutyl(3-ethoxy-5-methylphenyl)methanone ≥95%
Cyclobutyl(3-ethoxy-5-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 218.292 g/mol
MW: C₁₄H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3037676-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR