Cyclopropyl(3-ethoxy-5-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

446911-5G 446911-25G 446911-1G 446911-500MG
77629-352EA 1599.59 USD
77629-352 77635-838 77638-814 77650-816
Cyclopropyl(3-ethoxy-5-methylphenyl)methanone ≥95%
Cyclopropyl(3-ethoxy-5-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 204.265 g/mol
MW: C₁₃H₁₆O₂
Storage Temperature: Ambient
CAS Number: 3031808-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR