Cyclopropyl(3-ethoxy-5-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

446904-5G 446904-25G 446904-1G 446904-500MG
77629-348EA 1484.23 USD
77629-348 77635-836 77638-812 77650-814
Cyclopropyl(3-ethoxy-5-methylphenyl)methanol ≥95%
Cyclopropyl(3-ethoxy-5-methylphenyl)methanol

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031788-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR