1-(3-Ethoxy-5-methylphenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

446902-5G 446902-25G 446902-1G 446902-500MG
77629-346EA 1722.16 USD
77629-346 77655-766 77642-850 77646-844
1-(3-Ethoxy-5-methylphenyl)-2-methylpropan-1-one ≥95%
1-(3-Ethoxy-5-methylphenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031804-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR