1-(3-Ethoxy-5-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

446897-5G 446897-1G 446897-500MG
77655-764EA 1989 USD
77655-764 77642-848 77646-842
1-(3-Ethoxy-5-methylphenyl)propan-1-one ≥95%
1-(3-Ethoxy-5-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 192.254 g/mol
MW: C₁₂H₁₆O₂
Storage Temperature: Ambient
CAS Number: 1804281-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR