1-(3-Ethoxy-5-methylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

446887-5G 446887-25G 446887-1G 446887-500MG
77629-342EA 1484.23 USD
77629-342 77655-760 77642-844 77650-812
1-(3-Ethoxy-5-methylphenyl)propan-1-ol ≥95%
1-(3-Ethoxy-5-methylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 194.27 g/mol
MW: C₁₂H₁₈O₂
Storage Temperature: Ambient
CAS Number: 3031813-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR