(3-Ethoxy-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

446885-5G 446885-1G 446885-25G 446885-500MG
77633-158EA 1441 USD
77633-158 77638-810 77655-758 77646-838
(3-Ethoxy-5-methylphenyl)trimethylsilane ≥95%
(3-Ethoxy-5-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 208.372 g/mol
MW: C₁₂H₂₀OSi
Storage Temperature: Ambient
CAS Number: 3037676-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR