(3,5-Diethylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

446305-5G 446305-1G 446305-25G 446305-500MG
77633-156EA 1441 USD
77633-156 77638-806 77655-756 77650-808
(3,5-Diethylphenyl)(phenyl)methanol ≥95%
(3,5-Diethylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 240.34 g/mol
MW: C₁₇H₂₀O
Storage Temperature: Ambient
CAS Number: 3031789-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR