(3-Fluoro-2,4-dimethylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

446267-5G 446267-25G 446267-1G 446267-500MG
77633-154EA 1441 USD
77633-154 77659-350 77642-838 77646-836
(3-Fluoro-2,4-dimethylphenyl)(phenyl)methanol ≥95%
(3-Fluoro-2,4-dimethylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 230.277 g/mol
MW: C₁₅H₁₅FO
Storage Temperature: Ambient
CAS Number: 3031793-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR