1-(3-Fluoro-2,4-dimethylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

446248-5G 446248-1G 446248-25G 446248-500MG
77629-334EA 1484.23 USD
77629-334 77638-804 77659-348 77650-802
1-(3-Fluoro-2,4-dimethylphenyl)propan-1-ol ≥95%
1-(3-Fluoro-2,4-dimethylphenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 182.235 g/mol
MW: C₁₁H₁₅FO
Storage Temperature: Ambient
CAS Number: 3031808-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR