Ethyl(3-fluoro-2,4-dimethylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

446243-5G 446243-25G 446243-1G 446243-500MG
77629-332EA 1484.23 USD
77629-332 77635-832 77638-802 77650-800
Ethyl(3-fluoro-2,4-dimethylphenyl)sulfane ≥95%
Ethyl(3-fluoro-2,4-dimethylphenyl)sulphane

Precaución: For Research Use Only

Formula: 184.274 g/mol
MW: C₁₀H₁₃FS
Storage Temperature: Ambient
CAS Number: 3031804-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR