1-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

446145-5G 446145-1G 446145-25G 446145-500MG
77629-326EA 1484.23 USD
77629-326 77638-796 77655-752 77650-792
1-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-1-ol ≥95%
1-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-1-ol

Precaución: For Research Use Only

Formula: 234.2149 g/mol
MW: C₁₁H₁₃F₃O₂
Storage Temperature: Ambient
CAS Number: 3031804-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR