1-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

446140-5G 446140-1G 446140-500MG
77655-750EA 1898 USD
77655-750 77642-830 77650-788
1-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-1-one ≥95%
1-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 232.1990 g/mol
MW: C₁₁H₁₁F₃O₂
Storage Temperature: Ambient
CAS Number: 1261762-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR