α-Ethenyl-3-fluoro-5-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

445759-1G 445759-5G 445759-500MG
77638-792EA 537 USD
77638-792 77655-748 77646-830
α-Ethenyl-3-fluoro-5-(trifluoromethyl)benzenemethanol ≥95%
α-Ethenyl-3-fluoro-5-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 220.16 g/mol
MW: C₁₀H₈F₄O
Storage Temperature: Ambient
CAS Number: 1548384-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR