3-Chloro-α-ethenyl-4-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

445752-5G 445752-1G 445752-500MG
77629-322EA 1484.23 USD
77629-322 77642-828 77650-786
3-Chloro-α-ethenyl-4-(trifluoromethyl)benzenemethanol ≥95%
3-Chloro-α-ethenyl-4-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₈ClF₃O
Storage Temperature: Ambient
CAS Number: 2228659-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR