α-Cyclopropyl-4-methoxy-3-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

445339-5G 445339-1G 445339-500MG
77659-338EA 1898 USD
77659-338 77642-820 77650-780
α-Cyclopropyl-4-methoxy-3-(trifluoromethyl)benzenemethanol ≥95%
α-Cyclopropyl-4-methoxy-3-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: C₁₂H₁₃F₃O₂
Storage Temperature: Ambient
CAS Number: 2354303-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR