3-Chloro-α-cyclopropyl-4-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

445336-500MG 445336-5G 445336-1G
77652-820EA 379 USD
77652-820 77659-336 77642-818
3-Chloro-α-cyclopropyl-4-(trifluoromethyl)benzenemethanol ≥95%
3-Chloro-α-cyclopropyl-4-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 250.65 g/mol
MW: C₁₁H₁₀ClF₃O
Storage Temperature: Ambient
CAS Number: 2354614-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR