α-Cyclopropyl-2-fluoro-6-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

445330-5G 445330-25G 445330-1G 445330-500MG
77629-320EA 1484.23 USD
77629-320 77635-824 77642-816 77646-826
α-Cyclopropyl-2-fluoro-6-(trifluoromethyl)benzenemethanol ≥95%
α-Cyclopropyl-2-fluoro-6-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 234.19 g/mol
MW: C₁₁H₁₀F₄O
Storage Temperature: Ambient
CAS Number: 2832131-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR