3-Chloro-4'-iodo-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

444093-5G 444093-1G 444093-25G 444093-500MG
77629-318EA 1484.23 USD
77629-318 77638-790 77659-334 77646-824
3-Chloro-4'-iodo-1,1'-biphenyl ≥97%
3-Chloro-4'-iodo-1,1'-biphenyl

Precaución: For Research Use Only

Formula: 314.549 g/mol
MW: C₁₂H₈ClI
Storage Temperature: Ambient
CAS Number: 2439166-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR