1-(3'-Chloro-[1,1'-biphenyl]-4-yl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

444080-5G 444080-1G 444080-500MG
77659-332EA 1868 USD
77659-332 77646-822 77650-778
1-(3'-Chloro-[1,1'-biphenyl]-4-yl)-2-methylbutan-1-ol ≥95%
1-(3'-Chloro-[1,1'-biphenyl]-4-yl)-2-methylbutan-1-ol

Precaución: For Research Use Only

Formula: 274.785 g/mol
MW: C₁₇H₁₉ClO
Storage Temperature: Ambient
CAS Number: 3031808-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR