(3'-Chloro-[1,1'-biphenyl]-4-yl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

444075-5G 444075-500MG 444075-25G 444075-1G
77633-146EA 1441 USD
77633-146 77652-818 77655-742 77642-814
(3'-Chloro-[1,1'-biphenyl]-4-yl)trimethylsilane ≥95%
(3'-Chloro-[1,1'-biphenyl]-4-yl)trimethylsilane

Precaución: For Research Use Only

Formula: 260.834 g/mol
MW: C₁₅H₁₇ClSi
Storage Temperature: Ambient
CAS Number: 2439166-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR