(3'-Chloro-[1,1'-biphenyl]-4-yl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

444074-5G 444074-25G 444074-1G 444074-500MG
77633-144EA 1441 USD
77633-144 77659-330 77642-812 77646-820
(3'-Chloro-[1,1'-biphenyl]-4-yl)(methyl)sulfane ≥95%
(3'-Chloro-[1,1'-biphenyl]-4-yl)(methyl)sulphane

Precaución: For Research Use Only

Formula: C₁₃H₁₁ClS
Storage Temperature: Ambient
CAS Number: 844856-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR