(3'-Chloro-[1,1'-biphenyl]-4-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

444066-5G 444066-25G 444066-1G 444066-500MG
77633-142EA 618 USD
77633-142 77659-328 77642-810 77650-776
(3'-Chloro-[1,1'-biphenyl]-4-yl)boronic acid ≥95%
(3'-Chloro-[1,1'-biphenyl]-4-yl)boronic acid

Precaución: For Research Use Only

Formula: 232.471 g/mol
MW: C₁₂H₁₀BClO₂
Storage Temperature: Ambient
CAS Number: 1025496-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR