Ethyl 2-bromo-3,6-difluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

442693-5G 442693-25G 442693-1G 442693-500MG
77629-314EA 1484.23 USD
77629-314 77635-820 77642-808 77646-818
Ethyl 2-bromo-3,6-difluoro-α-oxobenzeneacetate ≥95%
Ethyl 2-bromo-3,6-difluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 293.06 g/mol
MW: C₁₀H₇BrF₂O₃
Storage Temperature: Ambient
CAS Number: 2004836-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR