Cyclopropyl(2,3-difluoro-4-pyridinyl)methanone ≥95%

Supplier: AOBChem USA

Warning

442601-5G 442601-1G 442601-500MG
77629-310EA 1484.23 USD
77629-310 77638-784 77650-774
Cyclopropyl(2,3-difluoro-4-pyridinyl)methanone ≥95%
Cyclopropyl(2,3-difluoro-4-pyridinyl)methanone

Precaución: For Research Use Only

Formula: 183.16 g/mol
MW: C₉H₇F₂NO
Storage Temperature: Ambient
CAS Number: 2353414-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR