Tert-butyl 2-(2-chloro-5-methylpyridin-3-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

439953-5G 439953-25G 439953-1G 439953-500MG
77629-308EA 1484.23 USD
77629-308 77635-818 77642-806 77646-814
Tert-butyl 2-(2-chloro-5-methylpyridin-3-yl)-2-oxoacetate ≥97%
Tert-butyl 2-(2-chloro-5-methylpyridin-3-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 255.6974 g/mol
MW: C₁₂H₁₄ClNO₃
Storage Temperature: Ambient
CAS Number: 3031789-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR