3,4-Difluoro-2-hydroxybenzonitrile ≥95%

Supplier: AOBChem USA

Warning

439709-5G 439709-1G 439709-500MG
77629-304EA 1484.23 USD
77629-304 77642-802 77646-808
3,4-Difluoro-2-hydroxybenzonitrile ≥95%
3,4-Difluoro-2-hydroxybenzonitrile

Precaución: For Research Use Only

Formula: C₇H₃F₂NO
Storage Temperature: Ambient
CAS Number: 186590-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR