5-Bromo-3-chloro-2-hydroxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

439704-5G 439704-1G 439704-500MG 439704-25G
77635-018EA 588 USD
77635-018 77638-778 77652-812 77655-734
5-Bromo-3-chloro-2-hydroxybenzonitrile ≥97%
5-Bromo-3-chloro-2-hydroxybenzonitrile

Precaución: For Research Use Only

Formula: C₇H₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD16249641
CAS Number: 1260810-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR