3-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

43834-5G 43834-25G 43834-1G 43834-500MG
77633-140EA 1168 USD
77633-140 77659-326 77642-800 77650-772
3-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥97%
3-Cyclopropyl-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

Precaución: For Research Use Only

Formula: 284.161 g/mol
MW: C₁₆H₂₁BN₂O₂
Storage Temperature: Ambient
CAS Number: 2377607-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR