3-(3,3-Dimethyl-1-butyn-1-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

436094-5G 436094-25G 436094-1G 436094-500MG
77633-138EA 1267 USD
77633-138 77659-324 77642-798 77650-770
3-(3,3-Dimethyl-1-butyn-1-yl)benzaldehyde ≥95%
3-(3,3-Dimethyl-1-butyn-1-yl)benzaldehyde

Precaución: For Research Use Only

Formula: C₁₃H₁₄O
MW: 186.25 g/mol
Storage Temperature: Ambient
CAS Number: 1392236-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR