Ethyl 4-cyclopropyl-2-oxo-3-butynoate ≥95%

Supplier: AOBChem USA

Warning

436074-5G 436074-25G 436074-1G 436074-500MG
77629-302EA 1484.23 USD
77629-302 77635-814 77642-796 77650-768
Ethyl 4-cyclopropyl-2-oxo-3-butynoate ≥95%
Ethyl 4-cyclopropyl-2-oxo-3-butynoate

Precaución: For Research Use Only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
CAS Number: 2244192-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR