1-(2-Methyl-3-(methylthio)phenyl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

436051-5G 436051-500MG 436051-25G 436051-1G
77629-300EA 1484.23 USD
77629-300 77652-810 77659-322 77642-794
1-(2-Methyl-3-(methylthio)phenyl)cyclopentanol ≥95%
1-(2-Methyl-3-(methylthio)phenyl)cyclopentanol

Precaución: For Research Use Only

Formula: 222.346 g/mol
MW: C₁₃H₁₈OS
Storage Temperature: Ambient
CAS Number: 3037705-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR