(2-Methyl-3-(methylthio)phenyl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

436021-5G 436021-25G 436021-1G 436021-500MG
77633-136EA 1441 USD
77633-136 77659-320 77642-790 77646-806
(2-Methyl-3-(methylthio)phenyl)(phenyl)methanone ≥95%
(2-Methyl-3-(methylthio)phenyl)(phenyl)methanone

Precaución: For Research Use Only

Formula: C₁₅H₁₄OS
Storage Temperature: Ambient
CAS Number: 3037693-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR