(4-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

435806-5G 435806-25G 435806-1G 435806-500MG
77633-134EA 1441 USD
77633-134 77659-316 77642-784 77646-802
(4-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl)trimethylsilane ≥95%
(4-Bromo-2-fluoro-3-(trifluoromethoxy)phenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 331.177 g/mol
MW: C₁₀H₁₁BrF₄OSi
Storage Temperature: Ambient
CAS Number: 3037705-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR