(2-Fluoro-3-(trifluoromethoxy)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

435805-5G 435805-25G 435805-1G 435805-500MG
77633-132EA 989 USD
77633-132 77655-726 77642-782 77650-756
(2-Fluoro-3-(trifluoromethoxy)phenyl)trimethylsilane ≥97%
(2-Fluoro-3-(trifluoromethoxy)phenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 252.281 g/mol
MW: C₁₀H₁₂F₄OSi
Storage Temperature: Ambient
CAS Number: 1878559-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR