1-(5-Methyl-2-thienyl)-1-propanone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(5-Methylthiophen-2-yl)propan-1-one

435431-5G 435431-25G 435431-1G 435431-500MG
77635-016EA 369 USD
77635-016 77659-312 77642-780 77650-752
1-(5-Methyl-2-thienyl)-1-propanone ≥97%
1-(5-Methyl-2-thienyl)-1-propanone

Precaución: For Research Use Only

Formula: C₈H₁₀OS
MW: 154.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00053143
CAS Number: 59303-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR