Trimethyl(5-methylthiophen-2-yl)silane ≥97%

Supplier: AOBChem USA

Warning

435430-5G 435430-1G 435430-500MG
77659-310EA 2250 USD
77659-310 77642-778 77650-750
Trimethyl(5-methylthiophen-2-yl)silane ≥97%
Trimethyl(5-methylthiophen-2-yl)silane

Precaución: For Research Use Only

Formula: 170.3472 g/mol
MW: C₈H₁₄SSi
Storage Temperature: Ambient
CAS Number: 18387-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR