1-(4-Methylthiophen-2-yl)butan-1-one ≥97%

Supplier: AOBChem USA

Warning

435428-1G 435428-5G 435428-500MG
77638-768EA 759 USD
77638-768 77655-724 77650-748
1-(4-Methylthiophen-2-yl)butan-1-one ≥97%
1-(4-Methylthiophen-2-yl)butan-1-one

Precaución: For Research Use Only

Formula: 168.2559 g/mol
MW: C₉H₁₂OS
Storage Temperature: Ambient
CAS Number: 501942-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR