1-(2-Isopropoxyphenyl)-2-(p-tolyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

435424-5G 435424-1G 435424-25G 435424-500MG
77629-286EA 1484.23 USD
77629-286 77638-766 77655-722 77646-800
1-(2-Isopropoxyphenyl)-2-(p-tolyl)ethanone ≥95%
1-(2-Isopropoxyphenyl)-2-(p-tolyl)ethanone

Precaución: For Research Use Only

Formula: 268.3502 g/mol
MW: C₁₈H₂₀O₂
Storage Temperature: Ambient
CAS Number: 1508157-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR