Methyl 2-methyl-2-(3-methylphenoxy)propanoate ≥95%

Supplier: AOBChem USA

Warning

434720-1G 434720-5G 434720-500MG
77638-764EA 574 USD
77638-764 77659-308 77650-746
Methyl 2-methyl-2-(3-methylphenoxy)propanoate ≥95%
Methyl 2-methyl-2-(3-methylphenoxy)propanoate

Precaución: For Research Use Only

Formula: 208.25 g/mol
MW: C₁₂H₁₆O₃
Storage Temperature: Ambient
CAS Number: 76410-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR