Methyl 2-(3-chlorophenoxy)-2-methylpropanoate ≥97%

Supplier: AOBChem USA

Warning

434706-5G 434706-1G 434706-500MG
77659-306EA 1911 USD
77659-306 77646-798 77650-744
Methyl 2-(3-chlorophenoxy)-2-methylpropanoate ≥97%
Methyl 2-(3-chlorophenoxy)-2-methylpropanoate

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₃
Storage Temperature: Ambient
CAS Number: 76410-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR