Ethyl 2-(isoquinolin-8-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

434441-500MG 434441-5G 434441-1G
77652-808EA 483 USD
77652-808 77659-304 77642-776
Ethyl 2-(isoquinolin-8-yl)-2-oxoacetate ≥95%
Ethyl 2-(isoquinolin-8-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₃H₁₁NO₃
Storage Temperature: Ambient
CAS Number: 380610-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR