1-((4-Fluorophenyl)ethynyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

434311-5G 434311-25G 434311-1G 434311-500MG
77633-126EA 1267 USD
77633-126 77659-302 77642-774 77650-742
1-((4-Fluorophenyl)ethynyl)cyclobutanol ≥95%
1-((4-Fluorophenyl)ethynyl)cyclobutanol

Precaución: For Research Use Only

Formula: C₁₂H₁₁FO
Storage Temperature: Ambient
CAS Number: 1394173-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR