3-(3-Fluorophenyl)-1-phenylprop-2-yn-1-ol ≥97%

Supplier: AOBChem USA

Warning

433764-5G 433764-1G 433764-25G 433764-500MG
77629-284EA 1484.23 USD
77629-284 77638-760 77655-718 77650-740
3-(3-Fluorophenyl)-1-phenylprop-2-yn-1-ol ≥97%
3-(3-Fluorophenyl)-1-phenylprop-2-yn-1-ol

Precaución: For Research Use Only

Formula: 226.2456 g/mol
MW: C₁₅H₁₁FO
Storage Temperature: Ambient
CAS Number: 1668554-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR