Ethyl 3-(1,1-dimethylethyl)-4-methoxy-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

433208-5G 433208-25G 433208-1G 433208-500MG
77629-278EA 1484.23 USD
77629-278 77635-804 77638-754 77650-734
Ethyl 3-(1,1-dimethylethyl)-4-methoxy-α-oxobenzeneacetate ≥95%
Ethyl 3-(1,1-dimethylethyl)-4-methoxy-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 264.32 g/mol
MW: C₁₅H₂₀O₄
Storage Temperature: Ambient
CAS Number: 1226332-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR