Ethyl 2-methoxy-3,5-dimethyl-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

433182-5G 433182-25G 433182-1G 433182-500MG
77629-276EA 1553.24 USD
77629-276 77635-802 77638-752 77646-790
Ethyl 2-methoxy-3,5-dimethyl-α-oxobenzeneacetate ≥95%
Ethyl 2-methoxy-3,5-dimethyl-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 236.26 g/mol
MW: C₁₃H₁₆O₄
Storage Temperature: Ambient
CAS Number: 1495548-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR