Methyl 3-bromo-2-fluoro-6-methoxy-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

432747-1G 432747-5G 432747-500MG
77638-748EA 628 USD
77638-748 77659-296 77650-730
Methyl 3-bromo-2-fluoro-6-methoxy-α-oxobenzeneacetate ≥95%
Methyl 3-bromo-2-fluoro-6-methoxy-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: 291.07 g/mol
MW: C₁₀H₈BrFO₄
Storage Temperature: Ambient
CAS Number: 2117246-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR