Methyl 3-chloro-2,6-difluoro-α-oxobenzeneacetate ≥95%

Supplier: AOBChem USA

Warning

432646-5G 432646-1G 432646-500MG
77659-294EA 2053 USD
77659-294 77642-766 77646-788
Methyl 3-chloro-2,6-difluoro-α-oxobenzeneacetate ≥95%
Methyl 3-chloro-2,6-difluoro-α-oxobenzeneacetate

Precaución: For Research Use Only

Formula: C₉H₅ClF₂O₃
Storage Temperature: Ambient
CAS Number: 2091025-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR